VWR® Disposable Tissue Grinders
Conical, 50 ml
Catalog # 47732-450
Supplier:   VWR International
VWR® Disposable Tissue Grinders
Catalog # 47732-450
Supplier:  VWR International
VWR® Disposable Tissue Grinders
VWR® Disposable Tissue Grinders
Conical, 50 ml
Price
$403.62
Log in pour la disponibilité
Case of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable tissue grinders are safe and easy to use.
  • Disposable
  • Safe and easy to use
  • Gamma sterilized
Their patented closed system has the ability to grind specimen in a sealed container, helping to prevent contact with the sample
Fully assembled and gamma sterilized. An extra cap and identification label are provided for storage, transport, and added safety. The 35 mL grinder (47732-448) is designed to be freestanding for easier handling.
The molded abrasive surface does not leave any sediment to obstruct examination of the sample.
Spécifications
  • Capacité:
    50 ml
  • Désignation:
    Conical