Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Caliper Electrodes, BTX | Avantor

Caliper Electrodes, BTX

Stainless Steel Caliper Electrode, 1.5 x 1.5 cm, 1 - 130 mm
Catalog # 47727-350
Supplier:   Harvard Apparatus
Fournisseur #:  45-0102
Caliper Electrodes, BTX
Catalog # 47727-350
Supplier:  Harvard Apparatus
Caliper Electrodes, BTX
Caliper Electrodes, BTX

Stainless Steel Caliper Electrode, 1.5 x 1.5 cm, 1 - 130 mm

Price
$2,282.97
Log in pour la disponibilité
Each
Product Details & Documents

In vivo-style, reusable electrodes are suitable for a variety of applications, including gene and transdermal drug delivery

Electrodes are compatible with electroporation systems 58018-004 and 58017-932. Electrodes complement Tweezertrodes™ 22221-518 and -520; Genetrodes® 47727-356 and -358; and 2-Needle Array™ electrode kits 22221-522 and -524.

Calipers may be cleaned with a mild detergent and sterilized with ethanol or ethylene oxide.

Each model consists of a caliper and a pair of end plates. End plates are adjusted using the black roller mounted on the caliper. Use brass caliper for smaller animals with a smaller surface area. Use stainless steel calipers for slightly larger surface areas.

Spécifications
  • Désignation:
    Stainless Steel Caliper Electrode
  • Dimensions:
    1.5 x 1.5 cm
  • Gap Size:
    1 - 130 mm
  • Cat. No.:
    47727-350
  • Voltage range:
    0 - 500 V
  • Pulse length range:
    1µs–99ms