Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
LPG Sample Cylinders, Koehler

LPG Sample Cylinders, Koehler

500 ml
Catalog # 37000-220
Supplier:   Koehler
Supplier #:  K27853
LPG Sample Cylinders, Koehler
Catalog # 37000-220
Supplier:  Koehler
LPG Sample Cylinders, Koehler
LPG Sample Cylinders, Koehler

500 ml

Price
$5,508.99
Log in for availability
Each
Product Details & Documents

Welded stainless steel cylinders obtain representative samples of liquefied petroleum gases (LPG)

Two-valve type instruments feature a 20% outage tube and built-in pressure relief valve preset between 540 and 600psi (38 and 42kg/cm2).

Certifications: Conform to specifications: ASTM D1265, GPA 2140.

Specifications
  • Capacité:
    500 ml