Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Drum Thieves, Koehler

Drum Thieves, Koehler

Stainless Steel, 710 ml (24 oz.), 32×1020 mm (1.25×40")
Catalog # 37000-186
Supplier:   Koehler
Supplier #:  K27401
Drum Thieves, Koehler
Catalog # 37000-186
Supplier:  Koehler
Drum Thieves, Koehler
Drum Thieves, Koehler

Stainless Steel, 710 ml (24 oz.), 32×1020 mm (1.25×40")

Price
$2,764.91
Log in for availability
Each
Product Details & Documents

Sampling tubes are suitable for taking bottom or other-level samples from barrels and drums.

Shipping weight: 2.7 kg (6 lbs.)

Specifications
  • Capacité:
    710 ml (24 oz.)
  • Désignation:
    Stainless Steel
  • O.D.×L:
    32×1020 mm (1.25×40")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.