Microscope Slide Labels, TimeMed
Thermal transfer labels, glove friendly, White, 22.2×22.2 mm (0.88×0.88"), 7400 labels/roll, 3 rolls (22200 labels)
36418-626
: TTR-7878
This collection of labels provides an assortment of instrument-specific applications for microscope slides.
The film MicroTrax labels (36418-606) are for use with MicroTrax thermal transfer printers and run four across on a roll. The standard thermal transfer labels (36418-626 and -628) are for use with most thermal transfer printers, and are supplied in rows of four on a roll. All labels are white.
- Color:White
- Désignation:Thermal transfer labels, glove friendly
- Dimensions:22.2×22.2 mm (0.88×0.88")
- Packaging:7400 labels/roll, 3 rolls (22200 labels)