Direct Thermal Printer Labels, TimeMed | Avantor
Direct Thermal Printer Labels, TimeMed
Permanent, coated thermal labels, White, 28.6×50.8 mm (1.13×2"), 5000 labels/roll, 6 rolls (30000 labels)
Catalog # 36418-608
Fournisseur #:  TD-PN7-S
Direct Thermal Printer Labels, TimeMed
Catalog # 36418-608
Direct Thermal Printer Labels, TimeMed
Direct Thermal Printer Labels, TimeMed
Permanent, coated thermal labels, White, 28.6×50.8 mm (1.13×2"), 5000 labels/roll, 6 rolls (30000 labels)
Price
$1,732.07
Log in pour la disponibilité
Case of 6
Product Details & Documents
This wide array of label types serves a variety of labeling applications in medical and research laboratories.
  • All labels are white unless otherwise noted
General-purpose labels are compatible with standard direct thermal printers. Printer-specific labels are designed for exclusive use with printers from the designated manufacturer.
Spécifications
  • Color:
    White
  • Désignation:
    Permanent, coated thermal labels
  • Dimensions:
    28.6×50.8 mm (1.13×2")
  • Packaging:
    5000 labels/roll, 6 rolls (30000 labels)