PYREX® Rectangular Chromatography Jars, Corning
11.4 L (3 gal.), 23.8 x 18.1 x 32.4 cm (93/8 x 71/8 x 123/4")
Supplier: Corning
Fournisseur #: 6944-11L
Jump to:
Product Details & Documents
Jars are ground to the close tolerance needed for a tight cover fit.
- These rectangular chromatography jars are made of Pyrex® borosilicate glass
Edges are ground flat within 0,25mm (0,01").
: Covers are not supplied.
: Not recommended for use with direct heat, pressure, or vacuum applications.
Spécifications
- Capacité:11.4 L (3 gal.)
- W×D×H:23.8 x 18.1 x 32.4 cm (93/8 x 71/8 x 123/4")



