Mantle/Controller Package, Series TM, Glas-Col®
3000 ml, 500 W
Catalog # 33752-500
Supplier: Glas-Col
: 100DTM112PL
Jump to:
Product Details & Documents
Glas-Col® offers TM series mantles and PowrTrol power control (see 33797-414 series) together in a special package
CSA certified.
The compact control is a solid-state, proportional voltage manual control that provides precise control of the output range, adjustable from 3–100%. Control has a lighted on/off power switch, three-wire load receptacle, and three-wire cord with molded plug. Series TM mantles feature an aluminum exterior with fabric interior, units up to 600W, 120V, 5A. Mantles withstand an internal operating temperature of 450°C (840°F).
Specifications
- Puissance:500 W
- For Flask Size:3000 ml