TuffStuff™ Critical Task Wipers, Contec® | Avantor
TuffStuff™ Critical Task Wipers, Contec®
TuffStuff™ heavy-duty wipes, quarter-folded, 30.5×43.2 cm (12×17"), 500/Bag, 2 Bags/Case
Catalog # 33503-134
Supplier:   CONTEC, INC. MX
Fournisseur #:  C1C1217Q
TuffStuff™ Critical Task Wipers, Contec®
Catalog # 33503-134
Supplier:  CONTEC, INC. MX
TuffStuff™ Critical Task Wipers, Contec®
TuffStuff™ Critical Task Wipers, Contec®
TuffStuff™ heavy-duty wipes, quarter-folded, 30.5×43.2 cm (12×17"), 500/Bag, 2 Bags/Case
Price
$393.88
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1,000
Product Details & Documents
TuffStuff™ Wipes are high-quality critical task wipes. Available in medium and heavy-duty formats, these wipes are solvent resistant and maintain their strength even when wet. TuffStuff™ wipes offer exceptional durability and cleanliness.
  • Hydroentangled nonwoven wipes are strong and durable without the use of binders
  • Medium and heavy-duty wipes available to match the ideal wipe for the specific application
  • Offers strength wet or dry, no residues or binders left behind
  • Highly sorbent without the use of surfactants
  • Multiple wipe sizes allow optimum selection for each job
TuffStuff™ Wipes are available in multiple sizes, weights, and packaging configurations to meet various customer needs. The expanded range includes TuffSat, consisting of heavy-duty dry wipes, quarter-folded in a tub that can be saturated with cleaning solution. This versatile option allows wipes to be customized with the correct cleaning solution for a particular environment.
Certifications: Recommended for ISO Class 6 to 8 environments.
Spécifications
  • Désignation:
    TuffStuff™ heavy-duty wipes, quarter-folded
  • Packaging:
    500/Bag, 2 Bags/Case
  • W×L:
    30.5×43.2 cm (12×17")