QuickConnect™ Mop Handles | Avantor
QuickConnect™ Mop Handles
QuickConnect™ anodized aluminum telescopic handle, 127 - 234 cm (50 - 92")
Catalog # 33502-715
Supplier:   Contec
Fournisseur #:  2643
QuickConnect™ Mop Handles
Catalog # 33502-715
Supplier:  Contec
QuickConnect™ Mop Handles
QuickConnect™ Mop Handles
QuickConnect™ anodized aluminum telescopic handle, 127 - 234 cm (50 - 92")
Price
$265.74
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Interchangeable mop handles are available in carbon fiber, anodized aluminum, stainless steel or fiberglass.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, anodized aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths. For hard-to-reach ceilings and walls, a carbon fiber handle is available that is lightweight and easy to maneuver. A one-piece stainless-steel handle is also available.
Spécifications
  • Désignation:
    QuickConnect™ anodized aluminum telescopic handle
  • Longueur:
    127 - 234 cm (50 - 92")