Polysoft Face Shields, OP-D-OP
Polysoft Face Shield, Blue
Catalog # 33000-456
Supplier:   OP-D-OP
 :  206PVRB
Polysoft Face Shields, OP-D-OP
Catalog # 33000-456
Supplier:  OP-D-OP
Polysoft Face Shields, OP-D-OP
Polysoft Face Shield, Blue
Price
$145.61
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Face shield is designed for facial protection in medical facilities and laboratories.
  • Unbreakable polycarbonate shield can be replaced
  • Cinch cord can be adjusted for a secure fit
  • Crystal clear optics
  • Polysoft band for comfort
Fits over glasses and replaces goggles and masks.
Spécifications
  • Color:
    Blue
  • Désignation:
    Polysoft Face Shield