Polysoft Face Shields, OP-D-OP
Polysoft Face Shield, Blue
Catalog # 33000-456
Supplier: OP-D-OP
: 206PVRB
Jump to:
Product Details & Documents
Face shield is designed for facial protection in medical facilities and laboratories.
- Unbreakable polycarbonate shield can be replaced
- Cinch cord can be adjusted for a secure fit
- Crystal clear optics
- Polysoft band for comfort
Fits over glasses and replaces goggles and masks.
Spécifications
- Color:Blue
- Désignation:Polysoft Face Shield