PYREX® Thermometer Adapters, Corning
19/22, 57 mm
Supplier: Corning
Supplier #: 8821-19
PYREX® Thermometer Adapters, Corning
Catalog # 32645-334
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Thermometer adapters are designed with a [ST] ground joint at one end; the other end is tooled to accommodate a thermometer.
- Supplied with a rubber thermometer holder
Note: These adapter tubes also function as a replacement part for all Corning organic chemistry kits.
Spécifications
- ST Joint No.:19/22
- Longueur:57 mm
- Cat. No.:32645-334
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact