PYREX® Thermometer Adapters, Corning
PYREX® Thermometer Adapters, Corning
19/22, 57 mm
Catalog # 32645-334
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  8821-19
PYREX® Thermometer Adapters, Corning
Catalog # 32645-334
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Thermometer Adapters, Corning
PYREX® Thermometer Adapters, Corning
19/22, 57 mm
Price
$66.10
Log in pour la disponibilité
Case of 1
Product Details & Documents
Thermometer adapters are designed with a [ST] ground joint at one end; the other end is tooled to accommodate a thermometer.
  • Supplied with a rubber thermometer holder
Note: These adapter tubes also function as a replacement part for all Corning organic chemistry kits.
Spécifications
  • ST Joint No.:
    19/22
  • Longueur:
    57 mm
  • Cat. No.:
    32645-334
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact