PYREX® Hybridization Tubes, Corning
Hybridization Tubes, with Cap and O-Ring, 35×300 mm, PYREX®, 45, PP cap
: Corning
: 7995-300
PYREX® Hybridization Tubes, Corning
32645-030
: Corning
Product available on GSA Advantage®
PYREX® borosilicate glass, GL 45 orange plug-seal cap with silicone O-ring.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
These tubes with three different lenghths are used in hybridiatzion incubators with rotator.
An optional Corning® vented membrane cap is available for low temperature assays.
- Cap Type:PP cap
- Ø×L:35×300 mm
- Désignation:Hybridization Tubes, with Cap and O-Ring
- Matière:PYREX®
- GL Thread Size:45
- Cat. No.:32645-030
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



