Blower Transition Adapters, Labconco®
Blower Transition Adapter, 30.5 cm (12"), 1.8 kg (4 lbs.)
Catalog # 30186-002
Supplier: Labconco
Fournisseur #: 7003400
Jump to:
Product Details & Documents
Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers
Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.
Spécifications
- Désignation:Blower Transition Adapter
- Shipping Weight:1.8 kg (4 lbs.)
- Nominal Diameter:30.5 cm (12")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



