PYREX® Stopcocks Keys, Corning | Avantor
PYREX® Stopcocks Keys, Corning
Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size C, 8 mm, Straight bore PTFE stopcocks
Catalog # 29610-642
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  7681-8
PYREX® Stopcocks Keys, Corning
Catalog # 29610-642
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Stopcocks Keys, Corning
PYREX® Stopcocks Keys, Corning
Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size C, 8 mm, Straight bore PTFE stopcocks
Price
$356.00
Log in pour la disponibilité
Case of 1
Product Details & Documents
These are replacement PTFE stopcock plug assemblies with various diameter bores for PYREX® straight bore PTFE stopcocks.
Spécifications
  • Ø trou:
    8 mm
  • Désignation:
    Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size C
  • For:
    Straight bore PTFE stopcocks
  • Cat. No.:
    29610-642
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact