PYREX® Stopcocks Keys, Corning
Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size C, 8 mm, Straight bore PTFE stopcocks
Supplier: Corning
Fournisseur #: 7681-8
PYREX® Stopcocks Keys, Corning
Catalog # 29610-642
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
These are replacement PTFE stopcock plug assemblies with various diameter bores for PYREX® straight bore PTFE stopcocks.
Spécifications
- Ø trou:8 mm
- Désignation:Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size C
- For:Straight bore PTFE stopcocks
- Cat. No.:29610-642
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact