PYREX® Stopcocks Keys, Corning | Avantor
PYREX® Stopcocks Keys, Corning
Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size B, 6 mm, Straight bore PTFE stopcocks
Catalog # 29610-640
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  7681-6
PYREX® Stopcocks Keys, Corning
Catalog # 29610-640
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Stopcocks Keys, Corning
PYREX® Stopcocks Keys, Corning
Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size B, 6 mm, Straight bore PTFE stopcocks
Price
$318.00
Log in pour la disponibilité
Case of 1
Product Details & Documents
These are replacement PTFE stopcock plug assemblies with various diameter bores for PYREX® straight bore PTFE stopcocks.
Spécifications
  • Ø trou:
    6 mm
  • Désignation:
    Replacement PTFE product standard straight bore stopcock plug assembly, O-ring size B
  • For:
    Straight bore PTFE stopcocks
  • Cat. No.:
    29610-640
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact