Storage Microplates 384-well Polypropylene, Corning®
Product Details & Documents
These 384-well polypropylene plates and blocks are built to the industry standard footprint
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
They are automation-friendly and solvent-resistant. They feature round-, conical-, or v-bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking. Both plates and blocks are certified DNase- and RNase-free.
Ordering information: The 384-well plates are packaged 25 per pack, except for 46610-064 which is packaged 10 per pack. The 384-well blocks are packaged 5 per pack.
Spécifications
- Sterility:Sterile
- Well Volume:180 µL
- Color:Clear
- Désignation:Round-Bottom
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Sealing Mat
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



