Corning® Pipette Tips, Microvolume
0.5 - 10 µl, +, Naturel, 10 Racks of 96
Supplier: Corning
Fournisseur #: 4830
Jump to:
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Spécifications
- Volume:0.5 - 10 µl
- Tip Color:Naturel
- Packed:10 Racks of 96
- Sterile:+