Corning® Pipette Tips, Microvolume
0.1 - 10 µl, -, Naturel, Rack Packed
Supplier: Corning
Supplier #: 4826
Corning® Pipette Tips, Microvolume
Catalog # 29443-298
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Specifications
- Volume:0.1 - 10 µl
- Tip Color:Naturel
- Packed:Rack Packed
- Sterile:-
- Cat. No.:29443-298
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact