Transwell® Clear Inserts, Polyester (PET) Membrane
6, 4.67 cm², 24 mm, Polyester, 3.0 µm, 6/Plate
Supplier: Corning
: 3452
Transwell® Clear Inserts, Polyester (PET) Membrane
Catalog # 29442-076
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
- w Produce a cell culture environment that closely resembles an in vivo state
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
- Treated for optimal cell attachment
- Excellent visibility under phase contrast microscopy
- Supplied in multiple well plates
- Sterile
Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.
Spécifications
- Nb de puits:6
- Taille de pore:3.0 µm
- Packaging:6/Plate
- Culture Area:4.67 cm²
- Membrane Diameter:24 mm
- Matériau membranaire:Polyester
- Cat. No.:29442-076
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact



