PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
12 L, 11
Supplier: Corning
Supplier #: 4060-12L
Jump to:
Product Details & Documents
Manufactured from borosilicate glass.
- High thermal shock resistance
- Provide contamination free results
- High chemical durability
Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).
Specifications
- Capacité:12 L
- Stopper No.:11