PYREX® Extraction Thimbles, with Fritted Disc, Corning
Medium, 35×90 mm, 170-200 µm
Supplier: Corning
Fournisseur #: 33950-MEC
PYREX® Extraction Thimbles, with Fritted Disc, Corning
Catalog # 27743-120
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with fritted disc.
- Resistant to chemical attack
- Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.
Spécifications
- Porosité:170-200 µm
- Taille:Medium
- Diameter ×H:35×90 mm
- Cat. No.:27743-120
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.