PYREX® Extraction Thimbles, with Fritted Disc, Corning | Avantor
PYREX® Extraction Thimbles, with Fritted Disc, Corning
Medium, 35×90 mm, 170-200 µm
Catalog # 27743-120
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  33950-MEC
PYREX® Extraction Thimbles, with Fritted Disc, Corning
Catalog # 27743-120
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Extraction Thimbles, with Fritted Disc, Corning
PYREX® Extraction Thimbles, with Fritted Disc, Corning
Medium, 35×90 mm, 170-200 µm
Price
$162.00
Log in pour la disponibilité
Case of 1
Product Details & Documents
PYREX®, borosilicate glass, with fritted disc.
  • Resistant to chemical attack
  • Excellent temperature resistance
It is designed for use with soxhlet extractors. It is also useful for chemical applications where it is required to weigh the extracted material.
Spécifications
  • Porosité:
    170-200 µm
  • Taille:
    Medium
  • Diameter ×H:
    35×90 mm
  • Cat. No.:
    27743-120
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact