Corning® Coliform Water Test Sample Container, Corning
Coliform Water Test Sample Container without Thiosulfate Tablet, 120.00 ml, Oui
Supplier: Corning
Fournisseur #: 1705-100
Jump to:
Product Details & Documents
These graduated, sterile, cylindrical vials made from PP are used in testing for the presence of coliform bacteria, microbiological contaminants in drinking water.
- One-piece container with attached lid to reduce chance of contamination
- Locking arrow ensures sterility
- Leak tight when sealed properly
- Tie-down to protect from accidental opening
- With or without sodium thiosulfate tablet
Meets EPA requirements
Ordering information: Includes 100 containers per case. Comes with sample labels and instructions.
Documents
Spécifications
- Avec bouchon:Oui
- Capacité:120.00 ml
- Désignation:Coliform Water Test Sample Container without Thiosulfate Tablet