Pliers Decappers, Wheaton
13 mm
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 224372
Jump to:
Product Details & Documents
Pliers decappers are an economical and easy way to remove seals.
To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal. Constructed of steel with red plastic-coated handles.
Specifications
- Seal Size:13 mm