Pliers Decappers, Wheaton
Pliers Decappers, Wheaton
13 mm
Catalog # 26676-428
Supplier #:  224372
Pliers Decappers, Wheaton
Catalog # 26676-428
Pliers Decappers, Wheaton
Pliers Decappers, Wheaton
13 mm
Price
$221.35
New Customer Promotional Price
Log in for availability
Case of 1
Product Details & Documents
Pliers decappers are an economical and easy way to remove seals.
To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal. Constructed of steel with red plastic-coated handles.
Specifications
  • Seal Size:
    13 mm