Split Stoppers, Labconco®
Split Stopper, 13 mm Corkage
Catalog # 26676-251
Supplier: Labconco
Fournisseur #: 7576010
Jump to:
Product Details & Documents
Split Stoppers
Package of 100. Rubber stopper, when crimped with an aluminum seal, tightly seals Serum Bottle for lyophilization applications. For use with FreeZone Freeze Dry Systems. Stoppers are for serum bottles with 20 mm corkage or 13 mm corkage, as indicated.
Spécifications
- Désignation:Split Stopper, 13 mm Corkage



