Inlet Filter, Labconco®
Replacement Filter Cartridge
26671-516
: Labconco
: 1472500
This disposable filter prevents oil backstreaming from the vacuum pump and protects the pump from damage.
Filter life is approximately 1000 hours at 10 µm vacuum.
: Includes filter assembly and one filter cartridge.
- Désignation:Replacement Filter Cartridge