Inlet Filter, Labconco®
Replacement Filter Cartridge
  26671-516
 :   Labconco
 :  1472500
Inlet Filter, Labconco®
  26671-516
 :  Labconco
Inlet Filter, Labconco®
Inlet Filter, Labconco®
Replacement Filter Cartridge
$1,100.00
Each
This disposable filter prevents oil backstreaming from the vacuum pump and protects the pump from damage.
Filter life is approximately 1000 hours at 10 µm vacuum.
 : Includes filter assembly and one filter cartridge.
  • Désignation:
    Replacement Filter Cartridge