Barnstead Automatic Collection System, Thermo Scientific | Avantor
Barnstead Automatic Collection System, Thermo Scientific
Flexible Tubing Adapter Kit
Catalog # 26299-454
Fournisseur #:  440138
Barnstead Automatic Collection System, Thermo Scientific
Catalog # 26299-454
Barnstead Automatic Collection System, Thermo Scientific
Barnstead Automatic Collection System, Thermo Scientific
Flexible Tubing Adapter Kit
Price
482,07 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Designed to collect water from the Barnstead Mega-Pure™ stills and control the operation of the Barnstead Mega-Pure™ Automatic Systems.
For use with models MP-3A, MP-6A, and MP-11A (see series 26294-257)
Use the optional tubing adapter kit where wall space requires more compact system. The optional outlet adapter allows an automatic dishwasher to be connected to collection bottle outlet. Equipped with shut-off stopcock.
A fully self-tending system consisting of 45L PYREX® brand glass collection bottle, mounting platform with dispensing valve and automatic control unit. Built-in controls protect system against overflow, waste of cooling water and over-temperature operation. Pure water is drawn from collection bottle; when water level falls below 35L, control sensors go into operation turning water supply back on and turning on electrical power to boiler.
Ordering information: Supplied with collection bottle, but without still or demineralizer units.
Spécifications
  • Désignation:
    Flexible Tubing Adapter Kit
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Product Outlet Tube
    Product Outlet Tube
    26299-400
    Log in pour la disponibilité
    Case of 1 - 510,93 $