Scheibler Desiccators | Avantor
Scheibler Desiccators
Scheibler Desiccators, ID, mm: 150
Catalog # 25010-060
Supplier:   Variety Glass
Fournisseur #:  682-3
Scheibler Desiccators
Catalog # 25010-060
Supplier:  Variety Glass
Scheibler Desiccators
Scheibler Desiccators
Scheibler Desiccators, ID, mm: 150
Price
$236.77
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Made of clear glass with thick uniform walls and a heavy, broad flange. Cover with molded knob is ground to fit the flange perfectly. Porcelain plates sold separately.
Spécifications
  • Désignation:
    Scheibler Desiccators, ID, mm: 150