Pre-Cleaned Cubitainer® Containers
Level III Containers, 9.5 L (2.5 gal.)
Catalog # 23050-304
Supplier: Environmental Express
Fournisseur #: EPC3010
Jump to:
Product Details & Documents
These collapsible LDPE plastic containers have polypropylene closures and are convenient for space-saving and storage
Containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance.
Cap size: 38-400.
Spécifications
- Capacité:9.5 L (2.5 gal.)
- Désignation:Level III Containers