Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Precleaned VOA Vials, Environmental Express® | Avantor

Precleaned VOA Vials, Environmental Express®

Level I quality assured/preserved vials with 1:1 HCL 0.2 ml, 40.00 ml, Clear, Oui, Open top
Catalog # 23050-298
Fournisseur #:  APC1601
Precleaned VOA Vials, Environmental Express®
Catalog # 23050-298
Precleaned VOA Vials, Environmental Express®
Precleaned VOA Vials, Environmental Express®

Level I quality assured/preserved vials with 1:1 HCL 0.2 ml, 40.00 ml, Clear, Oui, Open top

Price
$668.04
Log in pour la disponibilité
Case of 72
Product Details & Documents

These volatile organic analysis (VOA) vials are made of borosilicate glass. Choose from an open-top cap (0.125" thick PTFE/silicone septa) or closed-top style cap in a variarty of capacities and clear or amber glass.

Preserved vials come pre-filled with common preservative reagents for convenience and safety.

Certifications: All containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody-sealed, lot number-labeled for traceability, and a Certificate of Analysis indicating that the containers received full EPA quality assurance treatment.

Accessories information: The accessory Foam Block Rack holds 72×40 ml vials. Ideal for storage and shipping 40 ml VOA vials.

Documents

Spécifications
  • Avec bouchon:
    Oui
  • Cap Type:
    Open top
  • Capacité:
    40.00 ml
  • Color:
    Clear
  • Désignation:
    Level I quality assured/preserved vials with 1:1 HCL 0.2 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.