Precleaned VOA Vials, Environmental Express®
Product Details & Documents
These volatile organic analysis (VOA) vials are made of borosilicate glass. Choose from an open-top cap (0.125" thick PTFE/silicone septa) or closed-top style cap in a variarty of capacities and clear or amber glass.
Preserved vials come pre-filled with common preservative reagents for convenience and safety.
Certifications: All containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody-sealed, lot number-labeled for traceability, and a Certificate of Analysis indicating that the containers received full EPA quality assurance treatment.
Informations sur les accessoires: The accessory Foam Block Rack holds 72×40 ml vials. Ideal for storage and shipping 40 ml VOA vials.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Avec bouchon:Oui
- Cap Type:Closed top
- Capacité:20.00 ml
- Color:Clear
- Désignation:Level III vials
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



