Operating Scissors, Integra™ Miltex® | Avantor
Operating Scissors, Integra™ Miltex®
Sharp/Sharp, 15.2 cm (6"), Straight
Catalog # 21909-282
Supplier:   Miltex
Fournisseur #:  5-7
Operating Scissors, Integra™ Miltex®
Catalog # 21909-282
Supplier:  Miltex
Operating Scissors, Integra™ Miltex®
Operating Scissors, Integra™ Miltex®
Sharp/Sharp, 15.2 cm (6"), Straight
Price
$208.54
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Standard pattern
Stainless steel
Spécifications
  • Désignation:
    Sharp/Sharp
  • Longueur:
    15.2 cm (6")
  • Blade Type:
    Straight