Triangles
Triangles, iron wire, 6.4 cm (2.5")
Catalog # 21750-096
: H-23870
Jump to:
Discontinued
Product Details & Documents
These triangles are made up of iron wire (16 gauge) galvanized with plain clay pipe stems.
Smooth pipe stem triangles protect glassware on hotplates and over burner flames.
Spécifications
- Désignation:Triangles, iron wire, 6.4 cm (2.5")