Optimize Back Pressure Regulators
10 psi back pressure regulator
Catalog # 21510-684
Supplier: Optimize
Fournisseur #: 10-06-00120
Jump to:
Product Details & Documents
Optimize flow-through back pressure regulators utilize an active mechanism to maintain steady back pressure across a range of mobile phase viscosities and flow rates.
- Flow-through design
- Utilizes an active mechanism to maintain steady back pressure across a range of conditions
- Prevent solvent decompression and bubble formation in the flow cell
- Reduces baseline noise and drift
- 100 µl internal volume
Placing one downstream from your detector will prevent solvent decompression and bubble formation in the flow cell, reducing baseline noise and drift. Each pre-set Optimize stainless steel Back Pressure Regulator offers a 100 µl internal volume and includes fittings for immediate connection out of the box.
Delivery information: Includes OPTI-LOK® LP Fittings.
Spécifications
- Désignation:10 psi back pressure regulator