VWR® Micro Burets, Class A
Micro Buret, Class A, 2 ml
Catalog # 17477-041
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BURET CLS A RESERVOIR CS1 2ML Catalog #17555-021
Product 17555-021 is similar to 17477-041 but manufacturer and specifications may vary.
Product Details & Documents
Borosilicate glass buret manufactured to Class A specifications for burets.
- Borosilicate glass fabrication
Spécifications
- Capacité:2 ml
- Désignation:Micro Buret, Class A