VWR® Micro Burets, Class A
Micro Buret, Class A, 1 ml
17477-020
Borosilicate glass buret manufactured to Class A specifications for burets.
- Borosilicate glass fabrication
- Capacité:1 ml
- Désignation:Micro Buret, Class A