Microlink® Screw Caps, WHEATON
PP/Silicone/PTFE liners, 38-430, PP, Black
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 240114
Microlink® Screw Caps, WHEATON
Catalog # 16299-216
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Jump to:
Product Details & Documents
These screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- Liner is ultrasonically welded to the cap to eliminate the possibility of glue contamination and bond failure common with autoclaving
- Available in convenient lab pack quantities as well as larger case packs
- Low-compression-set silicone ideal for the storage of samples for prolonged period of time over wide temperature ranges
- PTFE or polypropylene on the exposed face of the liner imparts an inert,chemical-resistant surface
- Open top caps provide access with a syringe
- Autoclavable
Documents
Certificates
Recherche par numéro de lotSpécifications
- Taille du bouchon:38-430
- Color:Black
- Désignation:PP/Silicone/PTFE liners
- Matière:PP
- Cat. No.:16299-216