Polypropylene Snap-Seal™ Sample Containers, Corning® | Avantor
Polypropylene Snap-Seal™ Sample Containers, Corning®
300 ml (10.1 oz.), Snap-Seal™ Sample Containers
Catalog # 16126-030
Supplier:   Corning
Fournisseur #:  1730-10
Polypropylene Snap-Seal™ Sample Containers, Corning®
Catalog # 16126-030
Supplier:  Corning
Polypropylene Snap-Seal™ Sample Containers, Corning®
Polypropylene Snap-Seal™ Sample Containers, Corning®
300 ml (10.1 oz.), Snap-Seal™ Sample Containers
Price
$167.45
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 100
Product Details & Documents
PP, disposable, translucent.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both millilitres and ounces
  • Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.
Spécifications
  • Capacité:
    300 ml (10.1 oz.)
  • Désignation:
    Snap-Seal™ Sample Containers