Polypropylene Snap-Seal™ Sample Containers, Corning® | Avantor
Polypropylene Snap-Seal™ Sample Containers, Corning®
45 ml (1.5 oz.), Snap-Seal™ Sample Containers
Catalog # 16126-014
Supplier:   Corning
Fournisseur #:  1730-2C
Polypropylene Snap-Seal™ Sample Containers, Corning®
Catalog # 16126-014
Supplier:  Corning
Polypropylene Snap-Seal™ Sample Containers, Corning®
Polypropylene Snap-Seal™ Sample Containers, Corning®
45 ml (1.5 oz.), Snap-Seal™ Sample Containers
Price
$330.90
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 400
Product Details & Documents
PP, disposable, translucent.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both millilitres and ounces
  • Rough source for marking
These wide mouth sample containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly.
Spécifications
  • Capacité:
    45 ml (1.5 oz.)
  • Désignation:
    Snap-Seal™ Sample Containers