Pellet Cups® Sample Cups, Chemplex®
PlastiCups™, 35 mm (Pellet)
Catalog # 15566-068
Supplier: Chemplex
Fournisseur #: 553
Jump to:
Product Details & Documents
Compressible, tapered sample cups for converting powdered sample materials into self-supported briquettes.
- Available in plastic or aluminum
- Aluminum cups are taller than plastic cups and capable of accepting greater volumes of sample materials
The briquetted powdered samples formed are durable for safe handling, firm and resistant to fissure development, and reinforced on all sides except for the surface to be analyzed.
Spécifications
- Ø:35 mm (Pellet)
- Désignation:PlastiCups™
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.