Pellet Cups® Sample Cups, Chemplex®
Aluminum Pellet Cups, 40 mm (Pellet)
Catalog # 15566-064
Supplier: Chemplex
Fournisseur #: 545
Jump to:
Product Details & Documents
Compressible, tapered sample cups for converting powdered sample materials into self-supported briquettes.
- Available in plastic or aluminum
- Aluminum cups are taller than plastic cups and capable of accepting greater volumes of sample materials
The briquetted powdered samples formed are durable for safe handling, firm and resistant to fissure development, and reinforced on all sides except for the surface to be analyzed.
Spécifications
- Ø:40 mm (Pellet)
- Désignation:Aluminum Pellet Cups