Pellet Cups® Sample Cups, Chemplex® | Avantor
Pellet Cups® Sample Cups, Chemplex®
Aluminum Pellet Cups, 35 mm (Pellet)
Catalog # 15566-062
Supplier:   Chemplex
Fournisseur #:  535
Pellet Cups® Sample Cups, Chemplex®
Catalog # 15566-062
Supplier:  Chemplex
Pellet Cups® Sample Cups, Chemplex®
Pellet Cups® Sample Cups, Chemplex®
Aluminum Pellet Cups, 35 mm (Pellet)
Price
$701.54
Log in pour la disponibilité
Pack of 600
Product Details & Documents
Compressible, tapered sample cups for converting powdered sample materials into self-supported briquettes.
  • Available in plastic or aluminum
  • Aluminum cups are taller than plastic cups and capable of accepting greater volumes of sample materials
The briquetted powdered samples formed are durable for safe handling, firm and resistant to fissure development, and reinforced on all sides except for the surface to be analyzed.
Spécifications
  • Ø:
    35 mm (Pellet)
  • Désignation:
    Aluminum Pellet Cups