VWR® Temperature Chart Recorders
Le produit 33500-508 est une alternative fonctionnelle à 15551-010 mais l'emballage, la quantité, le fabricant et/ou les spécifications peuvent varier.
Product Details & Documents
Charts, markers, and batteries are preloaded; simply pull a tab to begin recording.
- Traceable®
- Factory calibrated
- Tamper-proof seal
- Display °C and °F on the same chart
Charts are easy to read, duplicate, fax, scan, and file
Temperature chart recorders are for monitoring refrigerators, supply rooms, food storage, chemical warehouses, animal quarters, incubators, goods-in-transit, walk-in chambers, biological materials, and manufacturing plants. Single-use, disposable recorders display temperatures for 5, 10, or 40 days on a 91 cm (36") strip chart. A solid-state, battery-driven, quartz-crystal controlled motor turns the chart with an accuracy of better than ±1 second per day. The temperature sensor is bimetallic. Accuracy is ±1°C (2°F).
Each recorder is factory calibrated. A tamper-evident seal maintains the integrity of the unit and results. A four-part form on the front allows space to note start time and pertinent information. Shock-resistant ABS case is 13.3x9.8x6.4 cm (51/4x37/8x21/2").
They show temperature and time/day of occurrence.
: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
: Supplied with AA alkaline battery.
Spécifications
- Recording Time:40 days