Microlink® Screw Caps, WHEATON
PP/Silicone/PP liners, 38-430, PP, White
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 240235-01
Jump to:
Product Details & Documents
These screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- Liner is ultrasonically welded to the cap to eliminate the possibility of glue contamination and bond failure common with autoclaving
- Available in convenient lab pack quantities as well as larger case packs
- Low-compression-set silicone ideal for the storage of samples for prolonged period of time over wide temperature ranges
- PTFE or polypropylene on the exposed face of the liner imparts an inert,chemical-resistant surface
- Open top caps provide access with a syringe
- Autoclavable
Documents
Certificates
Recherche par numéro de lotSpécifications
- Taille du bouchon:38-430
- Color:White
- Désignation:PP/Silicone/PP liners
- Matière:PP
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.