700 System Premium Receiver Head Assemblies, Techcon
0.9 m (3")
Catalog # 14219-728
Supplier: Techcon Systems
Fournisseur #: 70303RHB
Jump to:
Product Details & Documents
Head assemblies ensure accident-proof dispensing and provide a secure and reliable link between the dispensing barrel and the controller
Complete dispensing control. Compatible with all industry-standard dispensing barrels. Designed to exact industry standards.
Spécifications
- Longueur Tubes:0.9 m (3")