PCR Plates, Sorenson BioScience
96, 0.2 ml, Natural
Catalog # 14005-710
Supplier: Sorenson Bioscience
: 26190
Jump to:
Product Details & Documents
UltraAmp 96- and 384-well PCR plates contains bold-printed alphanumerics and fit all leading thermal cyclers, DNA sequencing/detection instruments, and Real-Time PCR instruments.
- Tight manufacturing tolerances and SBS standard dimensions create plates ideal for automated processes
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
Spécifications
- Nb de puits:96
- Well Color:Natural
- Well Volume:0.2 ml