PCR Plates, Sorenson BioScience
96, 0.2 ml, Natural
Catalog # 14005-710
Supplier:   Sorenson Bioscience
 :  26190
PCR Plates, Sorenson BioScience
Catalog # 14005-710
PCR Plates, Sorenson BioScience
PCR Plates, Sorenson BioScience
96, 0.2 ml, Natural
Price
$529.27
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 100
Product Details & Documents
UltraAmp 96- and 384-well PCR plates contains bold-printed alphanumerics and fit all leading thermal cyclers, DNA sequencing/detection instruments, and Real-Time PCR instruments.
  • Tight manufacturing tolerances and SBS standard dimensions create plates ideal for automated processes
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
Spécifications
  • Nb de puits:
    96
  • Well Color:
    Natural
  • Well Volume:
    0.2 ml