Orion™ Basic Electrodes, Thermo Scientific
Refillable Combination pH Electrode, Glass Body
Catalog # 14004-294
Supplier: Thermo Fisher Scientific
Supplier #: 9142BN
Jump to:
Product Details & Documents
Perform basic pH measurements quickly and reliably with Orion™ Basic Electrodes, for use with Orion™ Russell meters.
- pH range of 0 to 14
Specifications
- Désignation:Refillable Combination pH Electrode
- Matière:Glass Body
- Cat. No.:14004-294
- Operating temperature range:0 to 80°C