Orion™ Basic Electrodes, Thermo Scientific
Low Maintenance Gel Combination Electrode, with Removable Bulb Guard, Epoxy Body
  14004-288
 :  9146BN
Orion™ Basic Electrodes, Thermo Scientific
  14004-288
Orion™ Basic Electrodes, Thermo Scientific
Orion™ Basic Electrodes, Thermo Scientific
Low Maintenance Gel Combination Electrode, with Removable Bulb Guard, Epoxy Body
$236.29
Each
Perform basic pH measurements quickly and reliably with Orion™ Basic Electrodes, for use with Orion™ Russell meters.
  • pH range of 0 to 14
  • Désignation:
    Low Maintenance Gel Combination Electrode, with Removable Bulb Guard
  • Matière:
    Epoxy Body
  • Cat. No.:
    14004-288
  • Operating temperature range:
    0 to 80°C