Project Timer, Sper Scientific
Project timer, certified, 130×64×20 mm (5×2⅞×⅞"), 71 g (2.5 oz.)
Catalog # 14003-160
Supplier: Sper Scientific
Supplier #: 810017C
Jump to:
Product Details & Documents
This long range digital countdown project timer allows you to set a target day for completion of a project up to 8 years in the future and the unique timer counts down to your deadline in days, hours, minutes, seconds and ¹/₁₀₀ seconds on a 12 digit LCD.
- Standard clock, timer, and stopwatch modes
- A tripod back enables the project timer to stand on a bench top
View either the target date or the actual count down. You can insert the name of your project into a special window which can be seen on the timer face.
Certifications: N.I.S.T. traceable certificate of compliance for 14003-160.
Ordering information: One AAA battery included.
Specifications
- Désignation:Project timer, certified
- W×H×D:130×64×20 mm (5×2⅞×⅞")
- Weight:71 g (2.5 oz.)