Pipette Tips, Sorenson BioScience
MicroReach, 100 - 1000 µl, Bleu
Catalog # 13503-464
Supplier: Sorenson Bioscience
Fournisseur #: 27120
Jump to:
Product Details & Documents
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.
Spécifications
- Volume:100 - 1000 µl
- Tip Color:Bleu
- Tip Style:MicroReach